Cyclopentene-1-boronic acid, 97%
Cyclopentene-1-boronic acid, 97%
Cyclopentene-1-boronic acid, 97%
Cyclopentene-1-boronic acid, 97%
Thermo Scientific Chemicals

Cyclopentene-1-boronic acid, 97%

CAS: 850036-28-1 | C5H9BO2 | 111.935 g/mol
Have Questions?
Catalog NumberQuantity
H52531.MD
also known as H52531-MD
250 mg
Catalog number H52531.MD
also known as H52531-MD
Price (EUR)
227,80
Online Exclusive
268,00
Save 40,20 (15%)
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
227,80
Online Exclusive
268,00
Save 40,20 (15%)
Each
Add to cart
Chemical Identifiers
CAS850036-28-1
IUPAC Name(cyclopent-1-en-1-yl)boronic acid
Molecular FormulaC5H9BO2
InChI KeyUZBHNSVUMGIKLU-UHFFFAOYSA-N
SMILESOB(O)C1=CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (chromatographic, GC or HPLC)> 96%
Appearance (Color)White to pale yellow
Proton NMRConforms to structure
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only