5-Bromo-2-hydroxybenzeneboronic acid, 96%
5-Bromo-2-hydroxybenzeneboronic acid, 96%
5-Bromo-2-hydroxybenzeneboronic acid, 96%
5-Bromo-2-hydroxybenzeneboronic acid, 96%
Thermo Scientific Chemicals

5-Bromo-2-hydroxybenzeneboronic acid, 96%

CAS: 89598-97-0 | C6H6BBrO3 | 216.825 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H52937.03
also known as H52937-03
Price (EUR)
226,00
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
226,00
Each
Add to cart
Chemical Identifiers
CAS89598-97-0
IUPAC Name(5-bromo-2-hydroxyphenyl)boronic acid
Molecular FormulaC6H6BBrO3
InChI KeyXSXTWLOHAGPBNO-UHFFFAOYSA-N
SMILESOB(O)C1=C(O)C=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow-brown
Purity≥95.0%
FormSolid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only