sec-Butyl methyl ether, 99%
sec-Butyl methyl ether, 99%
sec-Butyl methyl ether, 99%
Thermo Scientific Chemicals

sec-Butyl methyl ether, 99%

CAS: 6795-87-5 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
100 mL
Catalog number H53427.AC
also known as H53427-AC
Price (EUR)
146,00
Each
Quantity:
25 mL
Request bulk or custom format
Price (EUR)
146,00
Each
Chemical Identifiers
CAS6795-87-5
IUPAC Name2-methoxybutane
Molecular FormulaC5H12O
InChI KeyFVNIMHIOIXPIQT-UHFFFAOYNA-N
SMILESCCC(C)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to pale yellow
FormLiquid
Assay from Supplier's CofA≥98.5%
sec-Butyl methyl ether is used as a solvent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
sec-Butyl methyl ether is used as a solvent in organic synthesis.

Solubility
Miscible with water, alcohol, ether and acetone.

Notes
Incompatible with strong oxidizing agents. Avoid conditions like flames, sparks and direct sunlight.
RUO – Research Use Only

General References:

  1. Kamiguchi, S.; Seki, Y.; Satake, A.; Okumura, K.; Nagashima, S.; Chihara, T. Catalytic Cracking of Methyl tert-Butyl Ether to Isobutene over Brønsted and Lewis Acid Sites on Solid-state Molybdenum Sulfide Clusters with an Octahedral Metal Framework. J. Cluster Sci. 2015, 26 (3), 653-660.
  2. Badia, J. H.; Fité, C.; Bringué, R.; Ramírez, E.; Cunill, F. Byproducts formation in the ethyl tert-butyl ether (ETBE) synthesis reaction on macroreticular acid ion-exchange resins. Appl. Catal., A 2013, 468, 384-394..