1-Trimethylsilyl-1,4-pentadiyne, 98%
1-Trimethylsilyl-1,4-pentadiyne, 98%
1-Trimethylsilyl-1,4-pentadiyne, 98%
1-Trimethylsilyl-1,4-pentadiyne, 98%
Thermo Scientific Chemicals

1-Trimethylsilyl-1,4-pentadiyne, 98%

CAS: 71789-10-1 | C8H12Si | 136.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H53436.03
also known as H53436-03
Price (EUR)
35,79
Online Exclusive
42,10
Save 6,31 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
35,79
Online Exclusive
42,10
Save 6,31 (15%)
Each
Add to cart
Chemical Identifiers
CAS71789-10-1
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)97.5% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Notes
Incompatible with strong oxidizing agents and ammonia.
RUO – Research Use Only

General References:

  1. Rodríguez, A. R.; Spur, B. W. First total synthesis of 7(S),17(S)-Resolvin D5, a potent anti-inflammatory docosanoid. Tetrahedron Lett. 2005, 46 (21), 3623-3627
  2. Zheng, X.; Herberich, G. E. Borabenzene Derivatives. 33. 3,5-Dimethylborabenzene 1,3,4,5-Tetramethylimidazol-2-ylidene: The First Carbene Adduct of a Borabenzene. Organometallics 2000, 19 (19), 3751-3753.