2,6-Dimethylphenylmagnesium bromide, 0.5M in 2-MeTHF
2,6-Dimethylphenylmagnesium bromide, 0.5M in 2-MeTHF
2,6-Dimethylphenylmagnesium bromide, 0.5M in 2-MeTHF
Thermo Scientific Chemicals

2,6-Dimethylphenylmagnesium bromide, 0.5M in 2-MeTHF

CAS: 21450-64-6 | C8H9BrMg | 209.37 g/mol
Have Questions?
Catalog NumberQuantity
H54138.AE
also known as H54138-AE
100 mL
Catalog number H54138.AE
also known as H54138-AE
Price (EUR)
151,00
Each
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
151,00
Each
Specifications
CAS21450-64-6
Chemical Name or Material2,6-Dimethylphenylmagnesium bromide
Concentration or Composition (by Analyte or Components)0.5M in 2-MeTHF
DOT InformationTransport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
Formula Weight209.37
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only