1-(p-Toluenesulfonyl)imidazole, 98+%
1-(p-Toluenesulfonyl)imidazole, 98+%
1-(p-Toluenesulfonyl)imidazole, 98+%
Thermo Scientific Chemicals

1-(p-Toluenesulfonyl)imidazole, 98+%

CAS: 2232-08-8 | C10H10N2O2S | 222.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number H54191.22
also known as H54191-22
Price (EUR)
745,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
745,00
Each
Chemical Identifiers
CAS2232-08-8
IUPAC Name1-(4-methylbenzenesulfonyl)-1H-imidazole
Molecular FormulaC10H10N2O2S
InChI KeyYJYMYJRAQYREBT-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)S(=O)(=O)N1C=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only