3,5-Difluorobenzylmagnesium bromide, 0.25M in 2-MeTHF
3,5-Difluorobenzylmagnesium bromide, 0.25M in 2-MeTHF
3,5-Difluorobenzylmagnesium bromide, 0.25M in 2-MeTHF
Thermo Scientific Chemicals

3,5-Difluorobenzylmagnesium bromide, 0.25M in 2-MeTHF

CAS: 1417985-27-3 | C7H5BrF2Mg | 231.323 g/mol
Have Questions?
Catalog NumberQuantity
H54427.AE
also known as H54427-AE
100 mL
Catalog number H54427.AE
also known as H54427-AE
Price (EUR)
224,00
Each
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
224,00
Each
Specifications
CAS1417985-27-3
Chemical Name or Material3,5-Difluorobenzylmagnesium bromide
Concentration or Composition (by Analyte or Components)0.25M in 2-MeTHF
DOT InformationTransport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
Formula Weight231.32
View more
It is a green alternative to Tetrahydrofuran. is capable of producing high concentrations of bromo, benzyl and allyl Grignard reagents. 2-MeTHF quickly produces clean water phase splits without forming emulsions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a green alternative to Tetrahydrofuran. is capable of producing high concentrations of bromo, benzyl and allyl Grignard reagents. 2-MeTHF quickly produces clean water phase splits without forming emulsions.

Solubilit
RUO – Research Use Only