3,5-Di-tert-butylcatechol, 99%
3,5-Di-tert-butylcatechol, 99%
3,5-Di-tert-butylcatechol, 99%
Thermo Scientific Chemicals

3,5-Di-tert-butylcatechol, 99%

CAS: 1020-31-1 | C14H22O2 | 222.328 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H55207.06
also known as H55207-06
Price (EUR)
35,30
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
35,30
Each
Chemical Identifiers
CAS1020-31-1
IUPAC Name3,5-di-tert-butylbenzene-1,2-diol
Molecular FormulaC14H22O2
InChI KeyPJZLSMMERMMQBJ-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC(O)=C(O)C(=C1)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>98.5%
It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Insoluble in water.

Notes
Store at room temperature with proper ventilation. Keep away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Gabor Speier.; Jozsef Csihony.; Anne M. Whalen.; Cortlandt G. Pierpont. Studies on Aerobic Reactions of Ammonia/3,5-Di-tert-butylcatechol Schiff-Base Condensation Products with Copper, Copper(I), and Copper(II). Strong Copper(II)-Radical Ferromagnetic Exchange and Observations on a Unique N-N Coupling Reaction. Inorg. Chem. 1996, 35 (12),3519-3524.
  2. Brandan A. Borgias.; Stephen R. Cooper.; Yun Bai Koh.; Kenneth N. Raymond. Synthetic, structural, and physical studies of titanium complexes of catechol and 3,5-di-tert-butylcatechol. Inorg. Chem. 1984, 23 (8),1009-1016.