Specifications
CAS39793-31-2
Chemical Name or Material4H-Thieno[3,2-b]pyrrole-5-carboxylic acid
Formula Weight167.19
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more