D-Alanine methyl ester hydrochloride, 98%
D-Alanine methyl ester hydrochloride, 98%
D-Alanine methyl ester hydrochloride, 98%
Thermo Scientific Chemicals

D-Alanine methyl ester hydrochloride, 98%

CAS: 14316-06-4 | C4H10ClNO2 | 139.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number H62472.14
also known as H62472-14
Price (EUR)
134,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
134,00
Each
Chemical Identifiers
CAS14316-06-4
IUPAC Namehydrogen methyl 2-aminopropanoate chloride
Molecular FormulaC4H10ClNO2
InChI KeyIYUKFAFDFHZKPI-UHFFFAOYNA-N
SMILES[H+].[Cl-].COC(=O)C(C)N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)>97.5%
D-Alanine methyl ester hydrochloride is used as building block for the preparation of peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Alanine methyl ester hydrochloride is used as building block for the preparation of peptides.

Solubility
Soluble in Water 100 mg/ml.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Bing Shi Li; Jacky W Y Lam; Zhen-Qiang Yu; Ben Zhong Tang. Tunable helical assemblies of L-alanine methyl ester-containing polyphenylacetylene. Langmuir. 2012, 28 (13), 5770-5774.
  2. Erik Schwartz; Pavol Bodis; Matthieu Koepf; Jeroen J L M Cornelissen; Alan E Rowan; Sander Woutersen; Roeland J M Nolte. Self-trapped vibrational states in synthetic beta-sheet helices. Chemical Communications. 2009, 31 4675-4677.