D-Tyrosine methyl ester hydrochloride, 98%
D-Tyrosine methyl ester hydrochloride, 98%
D-Tyrosine methyl ester hydrochloride, 98%
Thermo Scientific Chemicals

D-Tyrosine methyl ester hydrochloride, 98%

CAS: 3728-20-9 | C10H14ClNO3 | 231.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H63630.06
also known as H63630-06
Price (EUR)
55,10
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
55,10
Each
Chemical Identifiers
CAS3728-20-9
IUPAC Namemethyl (2R)-2-amino-3-(4-hydroxyphenyl)propanoate hydrochloride
Molecular FormulaC10H14ClNO3
InChI KeyVXYFARNRGZWHTJ-MVRPGTNWNA-N
SMILESCl.COC(=O)[C@H](N)CC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Proton NMRConforms to structure
Melting Point190-200°C
FormPowder or crystalline powder
Assay from Suppliers CofA≥97.5% (HPLC)
D-Tyrosine methyl ester hydrochloride is an intermediate in the synthesis of oxybutynin chloride (O868525), an inhibitor of proliferation and supresses gene expression in bladder smooth muscle cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Tyrosine methyl ester hydrochloride is an intermediate in the synthesis of oxybutynin chloride (O868525), an inhibitor of proliferation and supresses gene expression in bladder smooth muscle cells.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. John M. Park; Stuart B. Bauer; Michael R. Freeman; Craig A. Peters. Oxybutynin Chloride Inhibits Proliferation And Suppresses Gene Expression In Bladder Smooth Muscle Cells. The Journal of Urology. 1999, 162(3), 1110-1114.
  2. Nikki Watson; Donald V Daniels; Anthony P D W Ford; Richard M Eglen; and Sharath S Hegdes. Comparative pharmacology of recombinant human M3 and M5 muscarinic receptors expressed in CHO-K1 cells. Br J Pharmacol. 1999, 127(2), 590-596.