4-(Trifluoromethoxy)phenyl isothiocyanate, 97%
4-(Trifluoromethoxy)phenyl isothiocyanate, 97%
4-(Trifluoromethoxy)phenyl isothiocyanate, 97%
Thermo Scientific Chemicals

4-(Trifluoromethoxy)phenyl isothiocyanate, 97%

CAS: 64285-95-6 | C8H4F3NOS | 219.181 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64013.06
also known as H64013-06
Price (EUR)
158,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
158,00
Each
Chemical Identifiers
CAS64285-95-6
IUPAC Name1-isothiocyanato-4-(trifluoromethoxy)benzene
Molecular FormulaC8H4F3NOS
InChI KeyJKSZUQPHKOPVHF-UHFFFAOYSA-N
SMILESFC(F)(F)OC1=CC=C(C=C1)N=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only