4-Bromo-2,6-difluorophenylacetic acid, 96%, Thermo Scientific™
4-Bromo-2,6-difluorophenylacetic acid, 96%, Thermo Scientific™
4-Bromo-2,6-difluorophenylacetic acid, 96%, Thermo Scientific™
Thermo Scientific™

4-Bromo-2,6-difluorophenylacetic acid, 96%, Thermo Scientific™

Have Questions?
Catalog NumberQuantity
H66231.03
also known as H66231-03
1 g
Catalog number H66231.03
also known as H66231-03
Price (EUR)
163,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
163,00
Each
Specifications
CAS537033-54-8
Chemical Name or Material4-Bromo-2,6-difluorophenylacetic acid
Density1.77 g/mL
Formula Weight251.02
Health Hazard 1H315-H319-H335
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
RUO – Research Use Only