iBlot™ 2 Transfer Stacks, PVDF, mini
Invitrogen™
iBlot™ 2 Transfer Stacks, PVDF, mini
iBlot 2 Transfer Stacks are used to transfer proteins using the iBlot 2 Gel Transfer Device. They are consumable stacks with an integrated pre-activated PVDF transfer membrane for dry blotting of proteins.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
IB24002X33 x 10 stacks
IB2400210 Stacks
Catalog number IB24002X3
Price (EUR)
598,00
Each
Quantity:
3 x 10 stacks
Price (EUR)
598,00
Each
iBlot 2 Transfer Stacks are used to transfer proteins using the iBlot 2 Gel Transfer Device. They are consumable stacks with an integrated pre-activated PVDF transfer membrane for dry blotting of proteins. Each iBlot 2 Transfer Stack contains a copper-coated electrode and appropriate cathode and anode buffers in the gel matrix to allow fast, reliable transfer of proteins. This mini-size stack (8 x 8 cm) can be used with one mini-size gel.
PVDF membrane
The PVDF membrane (0.2-μm pore size, low fluorescence) in this stack has higher binding capacity than nitrocellulose. It is pre-activated and ready for use without any pretreatment with alcohols. The membrane is compatible with commonly used detection methods such as staining, immunodetection, fluorescence, and radiolabeling. The proteins bind to the membrane due to hydrophobic interactions. The protein-binding capacity is 240 μg/cm2.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity3 x 10 stacks
Dimensions (LxW)8 x 8 cm
FormatTransfer Stack
MaterialPVDF
Pore Size0.2 μm
Product LineiBlot
Unit SizeEach
Contents & Storage
Store at room temperature.