Thermo Scientific Chemicals

Tris(hydroxymethyl)aminomethane, Electrophoresis Grade, 99.5%

Catalog number: J61016.36
500 g, Each
Thermo Scientific Chemicals

Tris(hydroxymethyl)aminomethane, Electrophoresis Grade, 99.5%

Catalog number: J61016.36
500 g, Each
Quantity
Catalog number: J61016.36
also known as J61016-36
Price (EUR)
Price: 134,00
Online price: 120,60
Your price:
Quantity
-

Chemical Identifiers

CAS
77-86-1
IUPAC Name
2-amino-2-(hydroxymethyl)propane-1,3-diol
Molecular Formula
C4H11NO3
InChI Key
LENZDBCJOHFCAS-UHFFFAOYSA-N
SMILES
NC(CO)(CO)CO
Trace Metals
Fe: 5ppm max.
Trace Metals
Pb: 5ppm max.
Insoluble material
0.005% max.
Trace Metals
As: 5ppm max.
Trace Metals
Cu: 5ppm max.

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text