L-Cystine, Cell Culture Reagent
L-Cystine, Cell Culture Reagent
L-Cystine, Cell Culture Reagent
L-Cystine, Cell Culture Reagent
Thermo Scientific Chemicals

L-Cystine, Cell Culture Reagent

CAS: 56-89-3 | C6H12N2O4S2 | 240.292 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
1 kg
Catalog number J61651.A1
also known as J61651-A1
Price (EUR)
452,20
Online Exclusive
532,00
Save 79,80 (15%)
Each
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (EUR)
452,20
Online Exclusive
532,00
Save 79,80 (15%)
Each
Add to cart
Chemical Identifiers
CAS56-89-3
IUPAC Name(2R)-2-amino-3-{[(2R)-2-amino-2-carboxyethyl]disulfanyl}propanoic acid
Molecular FormulaC6H12N2O4S2
InChI KeyLEVWYRKDKASIDU-IMJSIDKUSA-N
SMILESN[C@@H](CSSC[C@H](N)C(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
CommentMicrobial Count: ≤1000 cfu/g (total plate count)
Loss on Drying≤0.2%
Sulfated Ash≤0.1%
FormCrystals or crystalline powder
View more
L-cysteine acts as a major source of sulfur in cell culture. Cysteine has been used in pulse-chase experiments to study protein expression from an inducible promoter. L-Cysteine is used to supplement cell culture media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-cysteine acts as a major source of sulfur in cell culture. Cysteine has been used in pulse-chase experiments to study protein expression from an inducible promoter. L-Cysteine is used to supplement cell culture media.

Solubility
It is soluble in water and HCL.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from heat.
RUO – Research Use Only

General References:

  1. Palacin, M., et al. The molecular bases of cystinuria and lysinuric protein intolerance. Curr. Opin. Genet. Dev. 2001, 11, (3), 328-335.
  2. McBean, G. J.; Flynn, J. Molecular mechanisms of cystine transport. Biochem. Soc. Trans. 2001, 29, ( Pt 6), 717-722.