D-(+)-Cellotetraose, 95%
D-(+)-Cellotetraose, 95%
D-(+)-Cellotetraose, 95%
D-(+)-Cellotetraose, 95%
Thermo Scientific Chemicals

D-(+)-Cellotetraose, 95%

CAS: 38819-01-1 | C24H42O21 | 666.579 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
10 mg
25 mg
Catalog number J67583.LB0
also known as J67583-#0
Price (EUR)
177,00
Each
Add to cart
Quantity:
5 mg
Request bulk or custom format
Price (EUR)
177,00
Each
Add to cart
Chemical Identifiers
CAS38819-01-1
IUPAC Name(2S,3R,4S,5S,6R)-2-{[(2R,3S,4R,5R,6S)-6-{[(2R,3S,4R,5R,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxan-3-yl]oxy}-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC24H42O21
InChI KeyLUEWUZLMQUOBSB-YQGOCCRESA-N
SMILESOC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@H]4[C@H](O)[C@@H](O)C(O)O[C@@H]4CO)O[C@@H]3CO)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Appearance (Color)White to off-white
Purity≥94.0%
Derived from cellulose degradation. Used by cellulosic bacteria as an energy source. Useful for characterization and identification of oligosaccharide metabolizing enzymes such as endoglucanases and --glucosidases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Derived from cellulose degradation
  • Used by cellulosic bacteria as an energy source
  • Useful for characterization and identification of oligosaccharide metabolizing enzymes such as endoglucanases and _x001A_-glucosidases
RUO – Research Use Only