D-(+)-Cellohexaose, 95%, Thermo Scientific Chemicals
D-(+)-Cellohexaose, 95%, Thermo Scientific Chemicals
D-(+)-Cellohexaose, 95%, Thermo Scientific Chemicals
D-(+)-Cellohexaose, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D-(+)-Cellohexaose, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mg
2 mg
5 mg
Catalog number J67610.LB0
also known as J67610-#0
Price (EUR)
349,00
Each
Add to cart
Quantity:
5 mg
Request bulk or custom format
Price (EUR)
349,00
Each
Add to cart
Chemical Identifiers
CAS2478-35-5
IUPAC Name(2R,3R,4R,5R)-4-{[(2S,3R,4R,5S,6R)-5-{[(2S,3R,4R,5S,6R)-5-{[(2S,3R,4R,5S,6R)-5-{[(2S,3R,4R,5S,6R)-3,4-dihydroxy-6-(hydroxymethyl)-5-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxan-2-yl]oxy}-3,4-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-3,4-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-3,4-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2,3,5,6-tetrahydroxyhexanal
Molecular FormulaC36H62O31
InChI KeyDJMVHSOAUQHPSN-VXDFXQCISA-N
SMILESOC[C@@H](O)[C@@H](O[C@@H]1O[C@H](CO)[C@@H](O[C@@H]2O[C@H](CO)[C@@H](O[C@@H]3O[C@H](CO)[C@@H](O[C@@H]4O[C@H](CO)[C@@H](O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O)[C@H](O)[C@H]4O)[C@H](O)[C@H]3O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥94.0%
FormCrystals or powder or crystalline powder
Appearance (Color)White to pale cream or pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only