3-Hexyne, 99%
3-Hexyne, 99%
3-Hexyne, 99%
Thermo Scientific Chemicals

3-Hexyne, 99%

CAS: 928-49-4 | C6H10 | 82.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L02682.14
also known as L02682-14
Price (EUR)
114,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
114,00
Each
Chemical Identifiers
CAS928-49-4
IUPAC Namehex-3-yne
Molecular FormulaC6H10
InChI KeyDQQNMIPXXNPGCV-UHFFFAOYSA-N
SMILESCCC#CCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.4095-1.4125 @ 20?C
FormLiquid
3-Hexyne is used in the synthesis of fused hetero-hydropyridyl ligands bonded to the {Ru(p-cymene)} organometallic moiety by reacting with ruthenacycles. It was also used in the preparation of [4+2] cycloaddition product by reacting with borole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hexyne is used in the synthesis of fused hetero-hydropyridyl ligands bonded to the {Ru(p-cymene)} organometallic moiety by reacting with ruthenacycles. It was also used in the preparation of [4+2] cycloaddition product by reacting with borole.

Solubility
Not miscible in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed in a cool, dry and well ventilated place.
RUO – Research Use Only

General References:

  1. Luciano Cuesta; Tatiana Soler; Esteban P Urriolabeitia. Cycloruthenated complexes from imine-based heterocycles: synthesis, characterization, and reactivity toward alkynes.Chemistry: A European Journal. 2012, 18,(47), 15178-15189.
  2. Fang Ge; Gerald Kehr; Constantin G Daniliuc; Gerhard Erker. Borole formation by 1,1-carboboration. Journal of the American Chemical Society. 2014, 136,(1), 68-71.
  3. On heating under pressure with norbornene and CO in the presence of Dodecacarbonyl triruthenium, 10507, alkynes undergo aromatization to give condensed hydroquinones: Organometallics, 17, 766 (1998). For reaction scheme, see 2-Hexyne, B22405.