Ethyl hydrogen glutarate, 97%, Thermo Scientific Chemicals
Ethyl hydrogen glutarate, 97%, Thermo Scientific Chemicals
Ethyl hydrogen glutarate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl hydrogen glutarate, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
L03941.06
also known as L03941-06
5 g
Catalog number L03941.06
also known as L03941-06
Price (EUR)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1070-62-8
IUPAC Name5-ethoxy-5-oxopentanoic acid
Molecular FormulaC7H12O4
InChI KeyMYMNBFURSYZQBR-UHFFFAOYSA-N
SMILESCCOC(=O)CCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4340-1.4390 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only