Thermo Scientific Chemicals

Methoxycarbonylsulfenyl chloride, 95%, Thermo Scientific Chemicals

Catalog number: L04350.03
1 g, Each
Thermo Scientific Chemicals

Methoxycarbonylsulfenyl chloride, 95%, Thermo Scientific Chemicals

Catalog number: L04350.03
1 g, Each
Quantity
Catalog number: L04350.03
also known as L04350-03
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
26555-40-8
IUPAC Name
(chlorosulfanyl)(methoxy)methanone
Molecular Formula
C2H3ClO2S
InChI Key
TXJXPZVVSLAQOQ-UHFFFAOYSA-N
SMILES
COC(=O)SCl
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
>94.0%
Refractive Index
1.4795-1.4865 @ 20?C

Description

Methoxycarbonylsulfenyl chloride is used to prepare 3-[(methoxycarbonyl)dithio]-L-alanine by reacting with L-cysteine. Further, it is used in the synthesis of 3-2 pyridinyldithio propanoic acid hydrazide (PDPH). It serves as an important heterobifunctional crosslinker, which is a crosslinker used to make macromolecule-drug conjugates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methoxycarbonylsulfenyl chloride is used to prepare 3-[(methoxycarbonyl)dithio]-L-alanine by reacting with L-cysteine. Further, it is used in the synthesis of 3-2 pyridinyldithio propanoic acid hydrazide (PDPH). It serves as an important heterobifunctional crosslinker, which is a crosslinker used to make macromolecule-drug conjugates.

Solubility
Miscible with dichlormethane.

Notes
Moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Flammable liquids
    Category 3
    Skin Corrosion/Irritation
    Category 1
    Serious Eye Damage/Eye Irritation
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H314 - Causes severe skin burns and eye damage

    Physical Hazards

    H226 - Flammable liquid and vapor

    Precautionary Statements

    P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

    P280 - Wear eye protection/ face protection

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P310 - Immediately call a POISON CENTER or doctor/physician