3,5-Dihydroxybenzonitrile, 98%
3,5-Dihydroxybenzonitrile, 98%
3,5-Dihydroxybenzonitrile, 98%
3,5-Dihydroxybenzonitrile, 98%
Thermo Scientific Chemicals

3,5-Dihydroxybenzonitrile, 98%

CAS: 19179-36-3 | C7H5NO2 | 135.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L05642.06
also known as L05642-06
Price (EUR)
217,60
Online Exclusive
256,00
Save 38,40 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
217,60
Online Exclusive
256,00
Save 38,40 (15%)
Each
Add to cart
Chemical Identifiers
CAS19179-36-3
IUPAC Name3,5-dihydroxybenzonitrile
Molecular FormulaC7H5NO2
InChI KeyABHOEQJNEOMTEK-UHFFFAOYSA-N
SMILESOC1=CC(=CC(O)=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream to brown
Melting Point (clear melt)185-197°C
Assay (HPLC)≥97.5%
3,5-Dihydroxybenzonitrile is used in the preparation of 3-hydroxy-5-methoxybenzonitrile using potassium carbonate as a reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dihydroxybenzonitrile is used in the preparation of 3-hydroxy-5-methoxybenzonitrile using potassium carbonate as a reagent.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Hilmer, A. J.; Bellisario, D. O.; Shimizu, S.; McNicholas, T. P.; Wang, Q. H.; Speakman, S. A.; Strano, M. S. Formation of High-Aspect-Ratio Helical Nanorods via Chiral Self-Assembly of Fullerodendrimers. J. Phys. Chem. Lett. 2014, 5 (5), 929-934.
  2. Liu, G.; Li, Z.; Wu, D.; Xue, W.; Li, T.; Liu, S. H.; Yin, J. Dendritic [2]Rotaxanes: Synthesis, Characterization, and Properties. J. Org. Chem. 2014, 79 (2), 643-652.