Thermo Scientific Chemicals

8-Hydroxy-2-methylquinoline, 98%, Thermo Scientific Chemicals

Catalog number: L06989.36
500 g, Each
Thermo Scientific Chemicals

8-Hydroxy-2-methylquinoline, 98%, Thermo Scientific Chemicals

Catalog number: L06989.36
500 g, Each
Quantity
Catalog number: L06989.36
also known as L06989-36
Price (EUR)
Price: 140,00
Online price: 124,60
Your price:
Quantity
-

Chemical Identifiers

CAS
826-81-3
IUPAC Name
2-methylquinolin-8-ol
Molecular Formula
C10H9NO
InChI Key
NBYLBWHHTUWMER-UHFFFAOYSA-N
SMILES
CC1=CC=C2C=CC=C(O)C2=N1
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
69.0-75.0?C
Appearance (Color)
Pale cream to cream to brown
Assay (GC)
≥97.5%

Description

8-Hydroxy-2-methylquinoline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Hydroxy-2-methylquinoline is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text