Thermo Scientific Chemicals

3-Methyl-1-phenyl-2-butanone, 96%, Thermo Scientific Chemicals

Catalog number: L07176.06
5 g, Each
Thermo Scientific Chemicals

3-Methyl-1-phenyl-2-butanone, 96%, Thermo Scientific Chemicals

Catalog number: L07176.06
5 g, Each
Quantity
Catalog number: L07176.06
also known as L07176-06
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
2893-05-2
IUPAC Name
3-methyl-1-phenylbutan-2-one
Molecular Formula
C11H14O
InChI Key
ZATIMOAGKJEVGN-UHFFFAOYSA-N
SMILES
CC(C)C(=O)CC1=CC=CC=C1
Refractive Index
1.5010-1.5070 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥95.0%

Description

3-Methyl-1-phenyl-2-butanone is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-1-phenyl-2-butanone is used as pharmaceutical intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text