Sodium bis(trimethylsilyl)amide, 1M soln. in THF
Sodium bis(trimethylsilyl)amide, 1M soln. in THF
Sodium bis(trimethylsilyl)amide, 1M soln. in THF
Thermo Scientific Chemicals

Sodium bis(trimethylsilyl)amide, 1M soln. in THF

CAS: 1070-89-9 | C6H18NNaSi2 | 183.377 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number L08779.AP
also known as L08779-AP
Price (EUR)
262,00
Each
Quantity:
500 mL
Request bulk or custom format
Price (EUR)
262,00
Each
Chemical Identifiers
CAS1070-89-9
IUPAC Namesodium bis(trimethylsilyl)azanide
Molecular FormulaC6H18NNaSi2
InChI KeyWRIKHQLVHPKCJU-UHFFFAOYSA-N
SMILES[Na+].C[Si](C)(C)[N-][Si](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Formliquid
Assay (Non-aqueous acid-base Titration)0.90-1.10M (18.3% - 22.4%)
Comment1.0M is 20.4%
Appearance (Color)Clear to slightly turbid, yellow to orange/brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium bis(trimethylsilyl)amide is useful for the generation of carbenes, the amination of sulfonamides, and for the preparation of lanthanide complexes. It is significantly accelerated the polymerization of pheneylacetylene in conjunction with rhodium (I) catalysis. It is used to deprotonate ketones and esters to generate enolate derivatives, phosphonium salts, generating Wittig reagents.

Solubility
Soluble in benzene, toluene and ether. Reacts with water.

Notes
Moisture Sensitive. Incompatible materials are Oxidizing agents, Oxygen. Store away from water and oxidizing agents.
RUO – Research Use Only

General References:

  1. Mario Janjetovic; Annika M Träff; Tobias Ankner; Jenny Wettergren; Göran Hilmersson. Solvent dependent reductive defluorination of aliphatic C-F bonds employing Sm(HMDS)2.Chemical Communications.2013, 49 (18), 1826-1828.
  2. Hua Lu; Jianjun Cheng. Hexamethyldisilazane-mediated controlled polymerization of alpha-amino acid N-carboxyanhydrides.Journal of the American Chemical Society.2007, 129 (46), 14114-14115.