Methyl 3-cyano-4-methoxybenzoate, 97%
Methyl 3-cyano-4-methoxybenzoate, 97%
Methyl 3-cyano-4-methoxybenzoate, 97%
Thermo Scientific Chemicals

Methyl 3-cyano-4-methoxybenzoate, 97%

CAS: 25978-74-9 | C10H9NO3 | 191.186 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L11107.06
also known as L11107-06
Price (EUR)
195,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
195,00
Each
Chemical Identifiers
CAS25978-74-9
IUPAC Namemethyl 3-cyano-4-methoxybenzoate
Molecular FormulaC10H9NO3
InChI KeyRYJSFYBJYKFNCF-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(C#N)=C(OC)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (GC)≥96.0%
Appearance (Color)Pale yellow to yellow
Melting Point (clear melt)119.0-128.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only