1-Benzyl-5-phenylbarbituric acid, 98+%
1-Benzyl-5-phenylbarbituric acid, 98+%
1-Benzyl-5-phenylbarbituric acid, 98+%
Thermo Scientific Chemicals

1-Benzyl-5-phenylbarbituric acid, 98+%

CAS: 72846-00-5 | C17H14N2O3 | 294.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L12954.14
also known as L12954-14
Price (EUR)
56,30
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
56,30
Each
Chemical Identifiers
CAS72846-00-5
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Melting Point (clear melt)161.0-167.0?C
FormPowder
Appearance (Color)White to cream
Assay (HPLC)≥98.0%
1-Benzyl-5-phenylbarbituric acid is useful as a pharmaceutical intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only