Ammonium hydroxide, 28% NH3
Ammonium hydroxide, 28% NH3
Ammonium hydroxide, 28% NH3
Thermo Scientific Chemicals

Ammonium hydroxide, 28% NH3

CAS: 1336-21-6 | H5NO | 35.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1000 mL
2500 mL
Catalog number L13168.AU
also known as L13168-AU
Price (EUR)
32,90
Each
Quantity:
1000 mL
Request bulk or custom format
Price (EUR)
32,90
Each
Chemical Identifiers
CAS1336-21-6
IUPAC Nameamine hydrate
Molecular FormulaH5NO
InChI KeyVHUUQVKOLVNVRT-UHFFFAOYSA-N
SMILESN.O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless
FormLiquid
Assay (Aqueous acid-base Titration)>26.5 to <34.0%
Ammonium hydroxide is widely utilized as a leavening agent or acidity regulator in food production. It serves as a precursor to some alkyl amines and is also used in the tobacco industry for flavor enhancement and as a processing aid. During furniture making, it combines with tannic acid and is used to darken or stain wood by making it iron salts. In chemical laboratories, it used for qualitative inorganic analysis, as a complexant and as a base. It is used to clean gold, silve, and platinum jewelry. It is an active component of Tollens' reagent (consisting of a solution of silver nitrate and ammonia) and is used to determine the presence of aldehyde or alpha-hydroxy ketone functional groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium hydroxide is widely utilized as a leavening agent or acidity regulator in food production. It serves as a precursor to some alkyl amines and is also used in the tobacco industry for flavor enhancement and as a processing aid. During furniture making, it combines with tannic acid and is used to darken or stain wood by making it iron salts. In chemical laboratories, it used for qualitative inorganic analysis, as a complexant and as a base. It is used to clean gold, silve, and platinum jewelry. It is an active component of Tollens′ reagent (consisting of a solution of silver nitrate and ammonia) and is used to determine the presence of aldehyde or alpha-hydroxy ketone functional groups.

Solubility
Miscible with water.

Notes
Incompatible with copper, iron and zinc.
RUO – Research Use Only

General References:

  1. Gu, X.; Zhou, K.; Freng, Y.; Yao, J. The morphology control of zeolitic imidazolate framework by addition of amino acid L-histidine. Chem. Lett. 2015, 44 (8), 1080-1082.
  2. Wang, D.; Belharouak, I.; Ortega, L. H.; Zhang, X.; Xu, R.; Zhou, D.; Zhou, G.; Amin, K. Synthesis of high capacity cathodes for lithium-ion batteries by morphology-tailored hydroxide co-precipitation. J. Power Sources. 2015, 274, 451-457.