Polysorbate 80
Polysorbate 80
Polysorbate 80
Thermo Scientific Chemicals

Polysorbate 80

CAS: 9005-65-6 | (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 | 604.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2500 mL
Catalog number L13315.AP
Price (EUR)
52,90
Each
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (EUR)
52,90
Each
Add to cart
Chemical Identifiers
CAS9005-65-6
Molecular Formula(C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
InChI KeyHDTIFOGXOGLRCB-KTKRTIGZNA-N
SMILESCCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
MDL NumberMFCD00082107
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange
FormViscous liquid
Acid Value≤2.0 mg KOH/g
CommentSpecification differs for U.S. and non-U.S. material where indicated
Identification (FTIR)Conforms (non-U.S. specification)
View more
Used for solubilizing membrane proteins during isolation and purification.Polysorbate 80 is used as a surfactant in soaps, cosmetics, body washes, hair shampoos, hair loss treatments, lotions, creams, bath oils and in mouthwash in the health care industry. It serves as an emulsifier in foods such as ice cream and in the preparation of antiarrhythmic amiodarone. Further, it acts as a solubilizing agent and excipient in the pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only