4-Methoxy-2-methylphenylhydrazine hydrochloride, 96%
4-Methoxy-2-methylphenylhydrazine hydrochloride, 96%
4-Methoxy-2-methylphenylhydrazine hydrochloride, 96%
Thermo Scientific Chemicals

4-Methoxy-2-methylphenylhydrazine hydrochloride, 96%

CAS: 93048-16-9 | C8H13ClN2O | 188.655 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L14810.03
also known as L14810-03
Price (EUR)
34,50
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
34,50
Each
Chemical Identifiers
CAS93048-16-9
IUPAC Name(4-methoxy-2-methylphenyl)hydrazine hydrochloride
Molecular FormulaC8H13ClN2O
InChI KeyOIFKWIPSJGNUCC-UHFFFAOYSA-N
SMILESCl.COC1=CC(C)=C(NN)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration ex Chloride)≥95.0 to ≤105.0%
Appearance (Color)White to cream to pale brown or pink
FormPowder or granules
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only