Sodium bromoacetate, 98%
Sodium bromoacetate, 98%
Sodium bromoacetate, 98%
Thermo Scientific Chemicals

Sodium bromoacetate, 98%

CAS: 1068-52-6 | C2H2BrNaO2 | 160.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L14983.14
also known as L14983-14
Price (EUR)
114,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
114,00
Each
Chemical Identifiers
CAS1068-52-6
IUPAC Namesodium 2-bromoacetate
Molecular FormulaC2H2BrNaO2
InChI KeySESSOVUNEZQNBV-UHFFFAOYSA-M
SMILES[Na+].[O-]C(=O)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
It is an important raw material and intermediate used in organic synthesis, agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Qifeng Zhang; Suobo Zhang; Lei Dai; Xuesi Chen. Novel zwitterionic poly(arylene ether sulfone)s as antifouling membrane material. Journal of Membrane Science.2010, 349(1-2), 217-224.
  2. Michael T. Crimmins; Patrick J. McDougall; and Kyle A. Emmitte. A Convergent Coupling Strategy for the Formation of Polycyclic Ethers: Stereoselective Synthesis of the BCDE Fragment of Brevetoxin A. Org. Lett.2005, 7(18), 4033-4036.