Thermo Scientific Chemicals

4,4'-(Hexafluoroisopropylidene)diphthalic anhydride, 99%, Thermo Scientific Chemicals

Catalog number: L16150.06
5 g, Each
Thermo Scientific Chemicals

4,4'-(Hexafluoroisopropylidene)diphthalic anhydride, 99%, Thermo Scientific Chemicals

Catalog number: L16150.06
5 g, Each
Quantity
Catalog number: L16150.06
also known as L16150-06
Price (EUR)
Price: 66,90
Online price: 60,21
Your price:
Quantity
-

Chemical Identifiers

CAS
1107-00-2
IUPAC Name
5-[2-(1,3-dioxo-1,3-dihydro-2-benzofuran-5-yl)-1,1,1,3,3,3-hexafluoropropan-2-yl]-1,3-dihydro-2-benzofuran-1,3-dione
Molecular Formula
C19H6F6O6
InChI Key
QHHKLPCQTTWFSS-UHFFFAOYSA-N
SMILES
FC(F)(F)C(C1=CC=C2C(=O)OC(=O)C2=C1)(C1=CC=C2C(=O)OC(=O)C2=C1)C(F)(F)F
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
241-248?C
Appearance (Color)
White to cream
Form
Powder

Description

4,4'-(Hexafluoroisopropylidene) diphthalic anhydride is involved in the synthesis of polyamides derived from terphenylenes, which is used for gas separation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-(Hexafluoroisopropylidene) diphthalic anhydride is involved in the synthesis of polyamides derived from terphenylenes, which is used for gas separation.

Solubility
Miscible with water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Corrosion/Irritation
    Category 1
    Serious Eye Damage/Eye Irritation
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H314 - Causes severe skin burns and eye damage

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P330 + P331 - IF SWALLOWED: Rinse mouth. Do NOT induce vomiting

    P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P310 - Immediately call a POISON CENTER or doctor/physician