N,N-Diethyl(2-chloro-1,1,2-trifluoroethyl)amine, 90%
N,N-Diethyl(2-chloro-1,1,2-trifluoroethyl)amine, 90%
N,N-Diethyl(2-chloro-1,1,2-trifluoroethyl)amine, 90%
N,N-Diethyl(2-chloro-1,1,2-trifluoroethyl)amine, 90%
Thermo Scientific Chemicals

N,N-Diethyl(2-chloro-1,1,2-trifluoroethyl)amine, 90%

CAS: 357-83-5 | C6H11ClF3N | 189.606 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L17330.14
also known as L17330-14
Price (EUR)
110,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
110,00
Each
Add to cart
Chemical Identifiers
CAS357-83-5
IUPAC Name(2-chloro-1,1,2-trifluoroethyl)diethylamine
Molecular FormulaC6H11ClF3N
InChI KeyBDZHKUAKSMWSAJ-UHFFFAOYNA-N
SMILESCCN(CC)C(F)(F)C(F)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.3905-1.3985 @ 20?C
Appearance (Color)Clear pale yellow to yellow to brown
Assay (GC)≥88.0%
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only