Ethyl 5-methoxyindole-2-carboxylate, 98%
Ethyl 5-methoxyindole-2-carboxylate, 98%
Ethyl 5-methoxyindole-2-carboxylate, 98%
Thermo Scientific Chemicals

Ethyl 5-methoxyindole-2-carboxylate, 98%

CAS: 4792-58-9 | C12H13NO3 | 219.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L17596.03
also known as L17596-03
Price (EUR)
36,20
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
36,20
Each
Chemical Identifiers
CAS4792-58-9
IUPAC Nameethyl 5-methoxy-1H-indole-2-carboxylate
Molecular FormulaC12H13NO3
InChI KeyNPIUAXNFAUGNHP-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC2=CC(OC)=CC=C2N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark yellow to yellow to brown
FormCrystals or powder or crystalline powder or granules
Assay (GC)≥97.5%
Melting Point (clear melt)154.0-160.0?C
It is reactant for preparation of ligands with binding specificity towards the GABA receptor, sodium-dependent glucose co-transporter 2 (SGLT2) inhibitors for the management of hyperglycemia in diabetes, anticancer agents,integrase strand-transfer inhibitors (INSTIs),inhibitor of Proliferation of Colon Cancer Cells ,isomeridianin G as GSK-3? inhibitors, HIV-1 integrase inhibitors and inhibitors of mitogen activated protein kinase-activated protein kinase 2 (MK-2).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is reactant for preparation of ligands with binding specificity towards the GABA receptor, sodium-dependent glucose co-transporter 2 (SGLT2) inhibitors for the management of hyperglycemia in diabetes, anticancer agents,integrase strand-transfer inhibitors (INSTIs),inhibitor of Proliferation of Colon Cancer Cells ,isomeridianin G as GSK-3? inhibitors, HIV-1 integrase inhibitors and inhibitors of mitogen activated protein kinase-activated protein kinase 2 (MK-2).

Solubility
Insoluble in water.

Notes
Store at room temperature. Keep away from strong oxidizing agents. Ensure proper ventilation.
RUO – Research Use Only
Thierry Fouquet.; Laurence Charles.; David Ruch. Collision-induced cage folding of partially condensed polyhedral oligomeric silsesquioxanes (POSS) bearing naked and capped silanols.Rapid Commun. Mass Spectrom. 2014 , 28 (16),1849-1852 .