(R)-(-)-4-Methyl-2-pentanol, 99%
(R)-(-)-4-Methyl-2-pentanol, 99%
(R)-(-)-4-Methyl-2-pentanol, 99%
(R)-(-)-4-Methyl-2-pentanol, 99%
Thermo Scientific Chemicals

(R)-(-)-4-Methyl-2-pentanol, 99%

CAS: 16404-54-9 | C6H14O | 102.177 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L18881.MD
also known as L18881-MD
Price (EUR)
49,80
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
49,80
Each
Add to cart
Chemical Identifiers
CAS16404-54-9
IUPAC Name4-methylpentan-2-ol
Molecular FormulaC6H14O
InChI KeyWVYWICLMDOOCFB-UHFFFAOYNA-N
SMILESCC(C)CC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Optical Rotation-21.0 ± 0.3? (neat)
FormLiquid
Refractive Index1.4100-1.4130 @ 20?C
Assay (GC)≥98.5%
For synthesis of optically active products

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only