2-Iodo-3-methylbenzoic acid, 98%
2-Iodo-3-methylbenzoic acid, 98%
2-Iodo-3-methylbenzoic acid, 98%
Thermo Scientific Chemicals

2-Iodo-3-methylbenzoic acid, 98%

CAS: 108078-14-4 | C8H7IO2 | 262.046 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L19876.06
also known as L19876-06
Price (EUR)
238,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
238,00
Each
Chemical Identifiers
CAS108078-14-4
IUPAC Name2-iodo-3-methylbenzoic acid
Molecular FormulaC8H7IO2
InChI KeyPZUXUOZSSYKAMX-UHFFFAOYSA-N
SMILESCC1=CC=CC(C(O)=O)=C1I
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)148.0-154.0?C
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only