Oxoid™ Tryptone Soya Yeast Extract Agar
Thermo Scientific™

Oxoid™ Tryptone Soya Yeast Extract Agar

Support the growth of a wide range of microorganisms with this agar, ideal for general microbial cultivation in laboratory settings.
Have Questions?
Catalog NumberQuantity
PO5050A10 Plates
Catalog number PO5050A
Price (EUR)
-
Quantity:
10 Plates
Support the growth of a wide range of microorganisms with Tryptone Soya Yeast Extract Agar, ideal for general microbial cultivation in laboratory settings.
Specifications
ColorStraw
Fill Weight17 g
Format90 mm Plate
Quantity10 Plates
Shelf Life182 days
Manufacturing OriginGermany
Product TypeAgar Plate
Unit SizeEach
Contents & Storage
2°C to 12°C