TriPlus™ RSH (SMART) Autosampler eLearning Courses
Thermo Scientific™

TriPlus™ RSH (SMART) Autosampler eLearning Courses

TriPLus™ RSH (SMART) Autosampler eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument. You will find a series of eLearning courses which can cover different topics such as foundational theory and/or basic operations principle and/or user maintenance and troubleshooting.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCourse Topic
TTPLRBOPBasic Operations
TTPLRMTSUser Maintenance and Troubleshooting
Catalog number TTPLRBOP
Price (EUR)
75,00
Each
Course Topic:
Basic Operations
Price (EUR)
75,00
Each

TriPLus™ RSH (SMART) Autosampler eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument. You will find a series of eLearning courses which can cover different topics such as foundational theory and/or basic operations principle and/or user maintenance and troubleshooting.

The TriPLus™ RSH (SMART) Autosampler basic operations with Chromeleon™ eLearning course
Familiarizes users with the basic sampling techniques offered on the Thermo Scientific™ TriPlus™ RSH and TriPlus™ RSH SMART autosampler. Topics covered in this course include:

  • Brief theory and the fundamentals of each sampling technique
  • Examples and applications of these techniques
  • Method creation in Chromeleon

The TriPlus™ RSH (SMART) Autosampler User Maintenance and Troubleshooting eLearning course is designed to provide you with the basic maintenance guidelines for the Thermo Scientific™ TriPlus™ RSH Autosamplers, including the following sections:

Maintenance menu section, which highlights the terminal functionality and provides basic maintenance checks recommended to avoid downtime.

Routine maintenance section, describing the typical routine maintenance that can be done by the user to keep the instrument functioning optimally.

The Teaching Modules section, which shows users how to teach modules in the event of a misalignment or if a module is added and needs to be taught.

Specifications
Access Availablity30 days upon end-user first access to the course
Course TopicBasic Operations
FormatOnline Self-paced eLearning
LanguagesEnglish
TechniqueGas Chromatography
Time and DateOn-demand upon course end-user registration confirmation
Course LevelFoundation
Duration<30 min.
For Use With (Equipment)TriPLus RSH (SMART) Autosampler
Product LineThermo Scientific
Product TypeChromatography
Target AudienceNew user seeking training on instrument operation and basic experimentation
Training LocationDigital access through Online Learning Management System
Unit SizeEach

Frequently asked questions (FAQs)

How do I purchase eLearning courses online with a purchase order (PO)?

To purchase an eLearning course with a PO, you need to have a purchasing account on thermofisher.com. For detailed steps on how to register and set up a purchasing account, please review this resource page (https://www.thermofisher.com/us/en/home/support/how-to-register-for-account.html). After your purchasing account has been set up, follow the instructions in this How-To-Guide (https://www.thermofisher.com/us/en/home/products-and-services/services/unity-lab-services/equipment-instrument-education/elearning-how-to-guide.html) for finding and ordering eLearning courses online with a PO.

How do I change to whom an eLearning course for analytical instruments and lab equipment is assigned?

Access Credit Manager to manage the eLearning course assignment. Select the eLearning course order and click on the Assign credits button to remove the existing user assignment.
Note: You can only change the assigned user if they have not yet accessed the eLearning course.

Can I assign an eLearning course for analytical instruments and lab equipment to more than one person?

You can assign an eLearning course for analytical instruments and lab equipment to more than one person only if you purchased multiple quantities. The number of quantities purchased equals the number of users that can be assigned to the course.

How long do I have to assign a purchased eLearning course for analytical instruments and lab equipment to a user?

You have one year from the date of purchase to assign an eLearning course for analytical instruments and lab equipment to a user.

What if the user I am delegating an eLearning course for analytical instruments and lab equipment to does not receive the email on how to manage eLearnings?

Please verify the email address used for delegating. If you are unable to locate the email, you can access Credit Manager directly.