Sulfato de magnesio heptahidrato, ACS, 98,0-102,0 %, Thermo Scientific Chemicals
Sulfato de magnesio heptahidrato, ACS, 98,0-102,0 %, Thermo Scientific Chemicals
Sulfato de magnesio heptahidrato, ACS, 98,0-102,0 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sulfato de magnesio heptahidrato, ACS, 98,0-102,0 %, Thermo Scientific Chemicals

Para protección contra incendios, textiles, portador de catalizador
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
100 g
500 g
2 kg
Número de catálogo 011596.36
también denominado 011596-36
Precio (EUR)
51,60
Each
Cantidad:
500 g
Pedido a granel o personalizado
Precio (EUR)
51,60
Each
Identificadores químicos
CAS10034-99-8
IUPAC Namemagnesium(2+) heptahydrate sulfate
Molecular FormulaH14MgO11S
InChI KeyWRUGWIBCXHJTDG-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Assay (unspecified)98.0-102.0% MgSO₄?7H₂O
Impurity contentNH₄ 0.002%
Impurity contentNO₃ 0.002%
Impurity contentSr 0.005%
pHof a 5% solution 5.0-8.2 at 25?
Ver más
Magnesium sulfate produced by appropriate process finds use in several pharmaceutical preparations of magnesium, usually known as epsom salt. It is used as a micronutrient in agriculture, brewing salt in production of certain alcoholic products, electrolyte to prepare metal salts, coagulant, and ingredient in bath salts for use in floatation therapy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only