Thermo Scientific Chemicals

Lámina delgada de níquel, 0,0025 mm de grosor, 99,95 % (base metálica), Thermo Scientific Chemicals

Número de catálogo: 040714.FI
50 x 50 mm, Each
Thermo Scientific Chemicals

Lámina delgada de níquel, 0,0025 mm de grosor, 99,95 % (base metálica), Thermo Scientific Chemicals

Número de catálogo: 040714.FI
50 x 50 mm, Each
Cantidad
Número de catálogo: 040714.FI
también denominado 040714-FI
Precio (EUR)
Precio: 346,00
Precio en línea: 307,94
Su precio:
Cantidad
-

Identificadores químicos

CAS
7440-02-0
IUPAC Name
nickel
Molecular Formula
Ni
InChI Key
PXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES
[Ni]
Appearance (Color)
Silvery white
Form
Foil, 0.0025mm (0.0001in) thick.
Assay from Supplier's CofA
≥99.95% (metals basis)

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text

    Seguridad y manipulación


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Sensitization
    Category 1
    Carcinogenicity
    Category 2
    Specific target organ toxicity - (repeated exposure)
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H317 - May cause an allergic skin reaction

    H351 - Suspected of causing cancer

    H372 - Causes damage to organs through prolonged or repeated exposure

    Precautionary Statements

    P201 - Obtain special instructions before use

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P308 + P313 - IF exposed or concerned: Get medical advice/attention

    P362 + P364 - Take off contaminated clothing and wash it before reuse