Ácido sulfúrico, sol. ac. 72 % p/p, Thermo Scientific Chemicals
Ácido sulfúrico, sol. ac. 72 % p/p, Thermo Scientific Chemicals
Ácido sulfúrico, sol. ac. 72 % p/p, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ácido sulfúrico, sol. ac. 72 % p/p, Thermo Scientific Chemicals

CAS: 7664-93-9 | H2O4S | 98.07 g/mol
Have Questions?
Número de catálogoCantidad
045596.K2
también denominado 045596-K2
1 l
Número de catálogo 045596.K2
también denominado 045596-K2
Precio (EUR)
162,00
Each
Cantidad:
1 l
Pedido a granel o personalizado
Precio (EUR)
162,00
Each
Identificadores químicos
CAS7664-93-9
IUPAC Namesulfuric acid
Molecular FormulaH2O4S
InChI KeyQAOWNCQODCNURD-UHFFFAOYSA-N
SMILESOS(O)(=O)=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay from Suppliers CofA72.0% w/w ± 3.6% w/w
Sulfuric acid has useful applications in domestic acidic drain cleaners, as a drying agent, as electrolytes in lead-acid batteries and various cleaning agents. It is also useful for mineral processing, fertilizer manufacture, oil refining, waste water processing and chemical synthesis. The sulfuric acid doped poly (m-aminophenol) is used as an aliphatic alcohol vapor sensor material. The catalytic decomposition of sulfuric acid on mixed Cr/Fe oxide shows useful application in sulfur-iodine cycle for hydrogen production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only