Thermo Scientific Chemicals

Bromuro de potasio, grado FTIR premium, Thermo Scientific Chemicals

Número de catálogo: 047111.KQ
200 Pc., Each
Thermo Scientific Chemicals

Bromuro de potasio, grado FTIR premium, Thermo Scientific Chemicals

Número de catálogo: 047111.KQ
200 Pc., Each
Cantidad
Número de catálogo: 047111.KQ
también denominado 047111-KQ
Precio (EUR)
Precio: 543,00
Precio en línea: 483,27
Su precio:
Cantidad
-

Identificadores químicos

CAS
7758-02-3
IUPAC Name
potassium bromide
Molecular Formula
BrK
InChI Key
IOLCXVTUBQKXJR-UHFFFAOYSA-M
SMILES
[K+].[Br-]
Form
Powder
Refractive Index
1.559

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text

    Seguridad y manipulación


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Serious Eye Damage/Eye Irritation
    Category 2
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H319 - Causes serious eye irritation

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing