Thermo Scientific Chemicals

Bismuth(III) selenide, 99.995% (metals basis), Thermo Scientific Chemicals

Número de catálogo: 047198.06
5 g, Each
Thermo Scientific Chemicals

Bismuth(III) selenide, 99.995% (metals basis), Thermo Scientific Chemicals

Número de catálogo: 047198.06
5 g, Each
Cantidad
Número de catálogo: 047198.06
también denominado 047198-06
Precio (EUR)
Cantidad
-

Identificadores químicos

CAS
12068-69-8
IUPAC Name
dibismuth(3+) triselandiide
Molecular Formula
Bi2Se3
InChI Key
FBGGJHZVZAAUKJ-UHFFFAOYSA-N
SMILES
[Se--].[Se--].[Se--].[Bi+3].[Bi+3]
Appearance (Color)
Black
Form
Powder
Total Metal Impurities
0.005% max.

Descripción

It is used as a semiconductor thermoelectric material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text

    Seguridad y manipulación


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 3
    Acute Inhalation Toxicity - Dusts and Mists
    Category 3
    Specific target organ toxicity - (repeated exposure)
    Category 2
    Acute aquatic toxicity
    Category 1
    Chronic aquatic toxicity
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H301 + H331 - Toxic if swallowed or if inhaled

    H373 - May cause damage to organs through prolonged or repeated exposure

    H410 - Very toxic to aquatic life with long lasting effects

    Precautionary Statements

    P260 - Do not breathe dust/fume/gas/mist/vapors/spray

    P264 - Wash face, hands and any exposed skin thoroughly after handling

    P301 + P310 - IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P311 - Call a POISON CENTER or doctor/physician

    P403 + P233 - Store in a well-ventilated place. Keep container tightly closed