Bromuro de etilmagnesio, 3 M en éter, Thermo Scientific Chemicals
Bromuro de etilmagnesio, 3 M en éter, Thermo Scientific Chemicals
Bromuro de etilmagnesio, 3 M en éter, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bromuro de etilmagnesio, 3 M en éter, Thermo Scientific Chemicals

Reactivo de Grignard
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 mol
0.5 mol
Número de catálogo 087291.JU
también denominado 087291-JU
Precio (EUR)
-
Cantidad:
1 mol
Pedido a granel o personalizado
Identificadores químicos
CAS925-90-6
IUPAC Namebromo(ethyl)magnesium
Molecular FormulaC2H5BrMg
InChI KeyTWTWFMUQSOFTRN-UHFFFAOYSA-M
SMILESCC[Mg]Br
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Clear to faintly turbid
FormLiquid
Concentration2.85-3.30M
Ethylmagnesium bromide is used as grignard reagent. It is widely used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only