Thermo Scientific Chemicals

N-Metilformanilida, 99+ %, Thermo Scientific Chemicals

Número de catálogo: 126810010
1 kg, Frasco de vidrio, Each
Thermo Scientific Chemicals

N-Metilformanilida, 99+ %, Thermo Scientific Chemicals

Número de catálogo: 126810010
1 kg, Frasco de vidrio, Each
Cantidad
Envase
Número de catálogo: 126810010
Precio (EUR)
Precio: 129,00
Precio en línea: 116,10
Su precio:
Cantidad
-

Identificadores químicos

CAS
93-61-8
IUPAC Name
N-methyl-N-phenylformamide
Molecular Formula
C8H9NO
InChI Key
JIKUXBYRTXDNIY-UHFFFAOYSA-N
SMILES
CN(C=O)C1=CC=CC=C1
GC
>=98.5 %
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless to yellow
Appearance (Form)
Liquid
Refractive index
1.5600 to 1.5620 (20°C, 589 nm)

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text

    Seguridad y manipulación


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 - Harmful if swallowed

    Precautionary Statements

    P264 - Wash face, hands and any exposed skin thoroughly after handling

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P312 - Call a POISON CENTER or doctor if you feel unwell