5-Cloro-2-pentanona etileno cetal, 97 %, Thermo Scientific Chemicals
5-Cloro-2-pentanona etileno cetal, 97 %, Thermo Scientific Chemicals
5-Cloro-2-pentanona etileno cetal, 97 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Cloro-2-pentanona etileno cetal, 97 %, Thermo Scientific Chemicals

CAS: 5978-08-5 | C7H13ClO2 | 164.63 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
5 g
25 g
Envase:
Frasco de vidrio
Número de catálogo 166040050
Precio (EUR)
52,10
Each
Cantidad:
5 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Precio (EUR)
52,10
Each
Identificadores químicos
CAS5978-08-5
IUPAC Name2-(3-chloropropyl)-2-methyl-1,3-dioxolane
Molecular FormulaC7H13ClO2
InChI KeyOFERIRWCHSOJJT-UHFFFAOYSA-N
SMILESCC1(CCCCl)OCCO1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=96.0 %
Refractive index1.4490 to 1.4510 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only