BOC-L-valina, 99+ %, Thermo Scientific Chemicals
BOC-L-valina, 99+ %, Thermo Scientific Chemicals
BOC-L-valina, 99+ %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

BOC-L-valina, 99+ %, Thermo Scientific Chemicals

CAS: 13734-41-3 | C10H19NO4 | 217.27 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
5 g
25 g
100 g
Envase:
Frasco de vidrio
Número de catálogo 275710050
Precio (EUR)
22,20
Each
Cantidad:
5 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Precio (EUR)
22,20
Each
Identificadores químicos
CAS13734-41-3
IUPAC Name(2S)-2-{[(tert-butoxy)carbonyl]amino}-3-methylbutanoic acid
Molecular FormulaC10H19NO4
InChI KeySZXBQTSZISFIAO-ZETCQYMHSA-N
SMILESCC(C)[C@H](NC(=O)OC(C)(C)C)C(O)=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Melting point76°C to 80°C
Infrared spectrumConforms
Specific optical rotation-5.3° to -7.3° (20°C, 589 nm) (c=1, CH3COOH)
Appearance (Form)Crystalline powder
HPLC>=99.0 %
Ver más
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only