Benzoilformato de etilo, 95 %, Thermo Scientific Chemicals
Benzoilformato de etilo, 95 %, Thermo Scientific Chemicals
Benzoilformato de etilo, 95 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzoilformato de etilo, 95 %, Thermo Scientific Chemicals

CAS: 1603-79-8 | C10H10O3 | 178.19 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
5 g
25 g
Envase:
Frasco de vidrio
Número de catálogo 410350250
Precio (EUR)
73,00
Each
Cantidad:
25 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Precio (EUR)
73,00
Each
Identificadores químicos
CAS1603-79-8
IUPAC Nameethyl 2-oxo-2-phenylacetate
Molecular FormulaC10H10O3
InChI KeyQKLCQKPAECHXCQ-UHFFFAOYSA-N
SMILESCCOC(=O)C(=O)C1=CC=CC=C1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
GC>=94 %
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.5150 to 1.5170 (20°C, 589 nm)
Appearance (Color)Clear colorless to green
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only