Acitretina
Thermo Scientific Chemicals

Acitretina

Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
5 g
Número de catálogo 466440050
Precio (EUR)
-
Cantidad:
5 g
Pedido a granel o personalizado
Identificadores químicos
CAS55079-83-9
IUPAC Name(2E,4E,6E,8E)-9-(4-methoxy-2,3,6-trimethylphenyl)-3,7-dimethylnona-2,4,6,8-tetraenoic acid
Molecular FormulaC21H26O3
InChI KeyIHUNBGSDBOWDMA-AQFIFDHZSA-N
SMILESCOC1=CC(C)=C(\C=C\C(\C)=C\C=C\C(\C)=C\C(O)=O)C(C)=C1C
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
HPLC>=98.0 %
Loss on drying=<0.5 %
Heavy metals=<20 ppm
Sulfated ash=<0.1 %
Appearance (Color)Yellow to green
Ver más
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only