Search Thermo Fisher Scientific
ε-Caprolactone is widely used as a monomer in the manufacturing of highly specialized polymers. It is mainly utilized as a precursor to caprolactam. It finds an application in the synthesis of polyglecaprone, which is used as a suture material in surgery.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Warning
H319 - Causes serious eye irritation
P264 - Wash face, hands and any exposed skin thoroughly after handling
P280 - Wear eye protection/ face protection
P337 + P313 - If eye irritation persists: Get medical advice/attention